missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenoxazine, 98%
CAS: 135-67-1 | C12H9NO | 183.21 g/mol
$333.64 - $333.64
Chemical Identifiers
| CAS | 135-67-1 |
|---|---|
| Molecular Formula | C12H9NO |
| Molecular Weight (g/mol) | 183.21 |
| MDL Number | MFCD00005014 |
| InChI Key | TZMSYXZUNZXBOL-UHFFFAOYSA-N |
| Synonym | phenoxazine, phenazoxine, unii-c2zwt499sg, c2zwt499sg, 5,4-oxazine, phenotazine, rarechem fh 2c 0s92, phenoxazine 1g, 9ci |
| PubChem CID | 67278 |
| IUPAC Name | 10H-phenoxazine |
| SMILES | N1C2=CC=CC=C2OC2=CC=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0196906
|
Thermo Scientific Chemicals
L0196906 |
5 g |
Each for $333.64
|
|
|||||
Description
Phenoxazine is used as primary and secondary intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenoxazine is used as primary and secondary intermediates.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Incompatible with oxidizing agents. Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances.
Chemical Identifiers
| 135-67-1 | |
| 183.21 | |
| TZMSYXZUNZXBOL-UHFFFAOYSA-N | |
| 67278 | |
| N1C2=CC=CC=C2OC2=CC=CC=C12 |
| C12H9NO | |
| MFCD00005014 | |
| phenoxazine, phenazoxine, unii-c2zwt499sg, c2zwt499sg, 5,4-oxazine, phenotazine, rarechem fh 2c 0s92, phenoxazine 1g, 9ci | |
| 10H-phenoxazine |
Specifications
| 135-67-1 | |
| C12H9NO | |
| 5 g | |
| 14,7253 | |
| Soluble in methanol. Insoluble in water. | |
| N1C2=CC=CC=C2OC2=CC=CC=C12 | |
| 183.21 | |
| 183.21 | |
| Phenoxazine |
| 154°C to 158°C | |
| MFCD00005014 | |
| 143234 | |
| phenoxazine, phenazoxine, unii-c2zwt499sg, c2zwt499sg, 5,4-oxazine, phenotazine, rarechem fh 2c 0s92, phenoxazine 1g, 9ci | |
| TZMSYXZUNZXBOL-UHFFFAOYSA-N | |
| 10H-phenoxazine | |
| 67278 | |
| 98% |
Safety and Handling
EINECSNumber : 205-210-8
RTECSNumber : SP7450000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only