missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenols Mix 1 Analytical Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Chemical Name or Material | Organic Pollutant Standards |
| Flash Point | 12°C |
| Packaging | Ampule |
| Quantity | 1 mL |
| UN Number | UN1219 - Class 3 - PG 2 - Isopropanol, solution |
| Grade | Analytical |