missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenolphthalein, U.S.P./N.F. grade, MP Biomedicals™
$80.75 - $80.75
Chemical Identifiers
| CAS | 77-09-8 |
|---|---|
| Molecular Formula | C20H14O4 |
| Molecular Weight (g/mol) | 318.33 |
| MDL Number | MFCD00005913 |
| InChI Key | KJFMBFZCATUALV-UHFFFAOYSA-N |
| Synonym | 3, 3-bis[4-Hydroxyphenyl]-1-[3H]- isobenzofuranone |
| PubChem CID | 4764 |
| ChEBI | CHEBI:34914 |
| IUPAC Name | 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one |
| SMILES | OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10260880
|
MP Biomedicals Inc
0210260880 |
100 g |
Each for $80.75
|
|
|||||
Chemical Identifiers
| 77-09-8 | |
| 318.33 | |
| KJFMBFZCATUALV-UHFFFAOYSA-N | |
| 4764 | |
| 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one |
| C20H14O4 | |
| MFCD00005913 | |
| 3, 3-bis[4-Hydroxyphenyl]-1-[3H]- isobenzofuranone | |
| CHEBI:34914 | |
| OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |
Specifications
| 77-09-8 | |
| C20H14O4 | |
| 3, 3-bis[4-Hydroxyphenyl]-1-[3H]- isobenzofuranone | |
| OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 | |
| 318.33 | |
| CHEBI:34914 | |
| 1.29g/cm3 | |
| 100 g | |
| Phenolphthalein |
| U.S.P./N.F. grade | |
| MFCD00005913 | |
| KJFMBFZCATUALV-UHFFFAOYSA-N | |
| 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one | |
| 4764 | |
| 318.3 | |
| White to White/Yellow | |
| Crystals |
Safety and Handling
EINECSNumber : 201-004-7
Recommended Storage : Store at 2° to 8°C.