missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein TS/RS, 1% (w/v) Alcoholic, Ricca Chemical

Catalog No. 562216
Change view
Click to view available options
Quantity:
250 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
562216 500 mL Natural Poly Bottle
R5622000250 250 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 562216 Supplier Ricca Chemical Company Supplier No. 562216
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Phenolphthalein TS/RS (USP)
CAS 64-17-5, 67-56-1, 77-09-8
Grade Indicator
Packaging Natural Poly Bottle
Color Colorless to Yellow
Quantity 500 mL