missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein 98.0+%, TCI America™

Catalog No. p-7134421
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P009425G 25 g
P0094500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P009425G Supplier TCI America Supplier No. P009425G
Only null left

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
Synonym phenolphthalein, phthalimetten, euchessina, phthalin, espotabs, phenolax, purgophen, koprol, laxogen, trilax
PubChem CID 4764
ChEBI CHEBI:34914
IUPAC Name 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Melting Point 262°C
Color White
Quantity 25 g
Formula Weight 318.33
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Phenolphthalein
EINECSNumber (9)-1152
RTECSNumber SM8380000
TSCA Yes