missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenolphthalein Stock Solution, Fisher Chemical™
Specifications
Specifications
| Chemical Name or Material | Phenolphthalein Stock Solution |
| CAS | 7732-18-5, 64-17-5, 67-56-1, 67-63-0, 77-09-8 |
| CAS Min % | 52.0, 43.0, 2.0, 2.0, 0.01 |
| CAS Max % | 55.0, 44.0, 3.0, 3.0, 0.01 |
| Packaging | Natural Poly Bottle |
| Color | Colorless |
| pH | 8 |
| Quantity | 50 mL |
| Grade | Indicator |