missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein StandAR, Macron Fine Chemicals™

Supplier:  Avantor H29505

Catalog No. MKH2951

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Phenolphthalein
77-09-8
C20H14O4
KJFMBFZCATUALV-UHFFFAOYSA-N
3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
4764
Glass
pH 8.0
1.0% in 95% Alcohol (APHA)
MFCD00005913
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
318.33
CHEBI:34914
1L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions