missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein Solution, Alcoholic 0.5%, Fisher Chemical

Catalog No. SP501
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SP501 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SP501 Supplier Thermo Fisher Scientific Supplier No. FLSP501
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Specifications

Chemical Name or Material Phenolphthalein Solution, Alcoholic 0.5%
Melting Point -89°C
Boiling Point 83°C
CAS 77-09-8, 7732-18-5
Concentration 0.45 to 0.55%
Packaging Glass Bottle
Assay Percent Range 0.5%
Vapor Pressure 40mmHg
Visual Transition Interval Pass Test
Density 0.7855g/cm³
Color Colorless
Quantity 1 L
Physical Form Liquid
Concentration or Composition (by Analyte or Components) 0.5%
Show More Show Less