missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein, Powder, Reagent, ACS, Spectrum™ Chemical

Catalog No. 18605095
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
50 kg
Packaging:
Poly Bottle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605095 6 x 500 g Poly Bottle
18605091 25 g Poly Bottle
18605092 125 g Poly Bottle
18605094 500 g Poly Bottle
18605093 6 x 125 g Poly Bottle
18605096 2.5 kg Poly Bottle
18605098 12 kg Poly Bottle
18605097 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18605095 Supplier Spectrum Chemical Mfg Cor Supplier No. P1075500GMCS6
Get Up to $500 in Chemicals with Purchase
View all promotions

P1075, 77-09-8, C20H14O4

Spectrum™ Chemical Phenolphthalein, Powder, Reagent, ACS is a chemical compound that varies in color according to the acidity level of the solution it is in. It has traditionally been used as a laxative but has more recently been used as part of a test to identify blood in substances, known as the Kastle-Mayer test. It is also used in certain toys to create disappearing ink effect. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
IUPAC Name 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 6 x 500 g
Grade Reagent
Physical Form Powder or Waxy Solid
Chemical Name or Material Phenolphthalein Powder