missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein Indicator, 1% (w/v) in Pyridine, Ricca Chemical

Catalog No. R5627000100
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R5627000100 100 mL Amber Glass Bottle
562716 500 mL Amber Glass Bottle
562732 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R5627000100 Supplier Ricca Chemical Company Supplier No. R5627000100C

Chemical Identifiers

CAS 110-86-1 , 77-09-8
Molecular Formula C5H5N
Molecular Weight (g/mol) Mixture
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

CAS Min % 97.0, 0.5
CAS Max % 100.0, 1.5
Concentration 1% (w/v)
Percent Purity 1% W/V
Grade Indicator
Packaging Amber Glass Bottle
Solvent or Matrix Pyridine
Color Colorless to Yellow
pH 8.5
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Phenolphthalein Indicator
Show More Show Less