missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein Indicator, 0.5% (w/v) in 50% (v/v) Alcohol, Neutralized, Ricca Chemical

Supplier:  Ricca Chemical Company R560000020F

 View more versions of this product

Catalog No. R560000020F


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Phenolphthalein Indicator
0.5% (w/v)
Indicator
50% (v/v) Alcohol
8
Liquid
7732-18-5,64-17-5,67-56-1,77-09-8
0.5% W/V
Cubitainer
Colorless to Pink
20 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions