missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein Indicator, 0.5% (w/v) in 50% (v/v) Alcohol, Neutralized, Ricca Chemical

Catalog No. 56004
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
56004 120 mL Natural Poly Bottle
560016 500 mL Natural Poly Bottle
560032 1 L Natural Poly Bottle
56001 4 L Natural Poly Bottle
R560000020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 56004 Supplier Ricca Chemical Company Supplier No. 56004

Specifications

Chemical Name or Material Phenolphthalein Indicator
CAS 7732-18-5, 64-17-5, 67-56-1, 77-09-8
Concentration 0.5% (w/v)
Percent Purity 0.5% W/V
Grade Indicator
Packaging Natural Poly Bottle
Solvent or Matrix 50% (v/v) Alcohol
Color Colorless to Pink
pH 8
Quantity 120 mL
Physical Form Liquid
Show More Show Less