missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenolphthalein Indicator, 0.5% (w/v) in 50% (v/v) Alcohol, Neutralized, Ricca Chemical
Supplier: Ricca Chemical Company 560032
Specifications
| Phenolphthalein Indicator | |
| 0.5% (w/v) | |
| Indicator | |
| 50% (v/v) Alcohol | |
| 8 | |
| Liquid |
| 7732-18-5,64-17-5,67-56-1,77-09-8 | |
| 0.5% W/V | |
| Natural Poly Bottle | |
| Colorless to Pink | |
| 1 L |