missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenolphthalein Indicator, 0.5% (w/v) in 50% (v/v) Alcohol, Neutralized, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Phenolphthalein Indicator |
| CAS | 7732-18-5, 64-17-5, 67-56-1, 77-09-8 |
| Concentration | 0.5% (w/v) |
| Percent Purity | 0.5% W/V |
| Grade | Indicator |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | 50% (v/v) Alcohol |
| Color | Colorless to Pink |
| pH | 8 |
| Quantity | 500 mL |
| Show More |