missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein Indicator, 0.08% (w/v) in Methanol, Ricca Chemical

Catalog No. 55954
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
55954 120 mL Natural Poly Bottle
559516 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 55954 Supplier Ricca Chemical Company Supplier No. 55954
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Phenolphthalein Indicator
CAS 67-56-1, 7732-18-5, 77-09-8
Percent Purity 0.08% W/V
Grade Indicator
Concentration or Composition (by Analyte or Components) 0.08% (w/v) in Methanol
Packaging Natural Poly Bottle
Color Colorless to Pink
Quantity 120 mL