missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein Indicator, 1% (w/v) in 95% (v/v) Alcohol, Neutralized, Ricca Chemical

Catalog No. 56204
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
56204 120 mL Natural Poly Bottle
562016 500 mL Natural Poly Bottle
562032 1 L Natural Poly Bottle
56201 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 56204 Supplier Ricca Chemical Company Supplier No. 56204
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Acid-Base Titrations

Specifications

Chemical Name or Material Phenolphthalein Indicator
CAS 64-17-5, 7732-18-5, 67-56-1, 77-09-8
Packaging Natural Poly Bottle
Solvent or Matrix 95% (v/v) Alcohol
Color Colorless to Pink
Quantity 120 mL
Percent Purity 1% W/V
Grade Indicator
Concentration or Composition (by Analyte or Components) 1% (w/v)