Learn More
Thermo Scientific Chemicals Phenolphthalein, ACS reagent
CAS: 77-09-8 | C20H14O4 | 318.33 g/mol
$91.00 - $297.32
Chemical Identifiers
| CAS | 77-09-8 |
|---|---|
| Molecular Formula | C20H14O4 |
| Molecular Weight (g/mol) | 318.33 |
| MDL Number | MFCD00005913 |
| InChI Key | KJFMBFZCATUALV-UHFFFAOYSA-N |
| PubChem CID | 4764 |
| ChEBI | CHEBI:34914 |
| SMILES | OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC417181000
|
Thermo Scientific Chemicals
417181000 |
100 g | Plastic bottle |
N/A
|
|
||||
|
AC417185000
|
Thermo Scientific Chemicals
417185000 |
500 g | Plastic bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
| 77-09-8 | |
| 318.33 | |
| KJFMBFZCATUALV-UHFFFAOYSA-N | |
| CHEBI:34914 |
| C20H14O4 | |
| MFCD00005913 | |
| 4764 | |
| OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |
Specifications
| 258.0°C to 263.0°C | |
| Passes Test | |
| MFCD00005913 | |
| 15, 7356 | |
| KJFMBFZCATUALV-UHFFFAOYSA-N | |
| OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 | |
| 318.33 | |
| CHEBI:34914 | |
| ACS Reagent | |
| Vapor Pressure: Negligible | |
| 1.299g/mL | |
| 100 g | |
| Phenolphthalein |
| 77-09-8 | |
| C20H14O4 | |
| 18, 143 | |
| Solubility in water: <0.1%. Other solubilities: 1g/12mL alcohol, 1g/100mL ether, soluble in alkalies, soluble in chloroform | |
| Authentic | |
| 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one | |
| 4764 | |
| 318.32 | |
| Plastic bottle | |
| from pH 8.0 (colorless) to pH 10 (red) | |
| White | |
| Crystalline Powder |
Safety and Handling
GHS H Statement:
May cause cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility.
GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Use personal protective equipment as required.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge
Danger
RUO – Research Use Only