missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein Disodium Salt (Water soluble), TCI America™

Catalog No. P009925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P009925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P009925G Supplier TCI America Supplier No. P009925G
Only null left

Chemical Identifiers

CAS 518-51-4
Molecular Formula C20H14O4
MDL Number MFCD00051414

Specifications

Quantity 25 g
Formula Weight 362.29
Physical Form Crystalline Powder
Chemical Name or Material Phenolphthalein Disodium Salt (Water soluble)