missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phenolphthalein, ACS reagent

Supplier:  Thermo Scientific™ 417180026

Catalog No. AC417180026


Only null left

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Indicator
Specifications

Specifications

Phenolphthalein
Passes Test
MFCD00005913
15, 7356
KJFMBFZCATUALV-UHFFFAOYSA-N
Authentic
318.33
CHEBI:34914
ACS Reagent
from pH 8.0 (colorless) to pH 10 (red)
White
77-09-8
C20H14O4
18, 143
Solubility in water: <0.1%. Other solubilities: 1g/12mL alcohol, 1g/100mL ether, soluble in alkalies, soluble in chloroform
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
4764
318.32
Plastic bottle
1.299g/mL
2.5kg
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility.

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

Danger

SDS
Documents

Documents

Promotions

Promotions