missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein, ACS Grade, LabChem™

Supplier:  LabChem LC181989

Catalog No. LC181989


Only null left

Description

Description

Specifications

Specifications

Phenolphthalein
450°C
100
C20H14O4
Soluble in water
KJFMBFZCATUALV-UHFFFAOYSA-N
3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
4764
318.32
Poly Bottle
Carbon dioxide; Carbon monoxide
White
Powder
262°C
77-09-8
C20H14O4
MFCD00005913
Passes Test
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
318.33
CHEBI:34914
ACS
1277kg/m3
1,277kg/m3
100 g

Chemical Identifiers

77-09-8
318.33
KJFMBFZCATUALV-UHFFFAOYSA-N
CHEBI:34914
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
C20H14O4
MFCD00005913
4764
3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing cancer.

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves/eye protection/face protection.
If exposed or concerned: Get medical attention.
Store locked up.
Dispose of contents/container in accordance with local, state and federal regulations.

Warning

EINECSNumber : 201-004-7

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions