missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenolphthalein, ACS

Indicator | CAS: 77-09-8 | C20H14O4 | 318.33 g/mol

Supplier:  Thermo Scientific Chemicals 3870522

 View more versions of this product

Catalog No. AA3870522


Only null left

Description

Description

Phenolphthalein is widely used as an indicator in acid-base titrations, water treatment applications. Due to its pH sensitivity, It is used to the check extend of carbonation in concrete, soap and in toys as diapering work. It is used in Kastle-Meyer test, which identifies the substances containing blood.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phenolphthalein
For analysis ACS
C20H14O4
284423
Soluble in alcohol; Slightly soluble in ether; Slightly soluble in dimethyl sulfoxide and insoluble in benzene or hexane.
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
318.33
CHEBI:34914
ACS
100 g
258°C to 262°C
77-09-8
MFCD00005913
14,7243
KJFMBFZCATUALV-UHFFFAOYSA-N
3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
4764
318.33
1.299 g/mL
Powder

Chemical Identifiers

77-09-8
318.33
KJFMBFZCATUALV-UHFFFAOYSA-N
CHEBI:34914
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
C20H14O4
MFCD00005913
4764
3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H341-H361

P201-P202-P260-P281-P308+P313-P501c

H341-H351-H361f-H373

EINECSNumber : 201-004-7

RTECSNumber : SM8380000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only