missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenolphthalein, 1% (w/v) Indicator Solution in 95% Isopropyl Alcohol, Spectrum™ Chemical
Specifications
Specifications
| Chemical Name or Material | Phenolphthalein |
| CAS | 77-09-8, 67-63-0, 7732-18-5 |
| Concentration | 1 Percent (w/v) Indicator Solution in 95 Percent Isopropyl Alcohol |
| Grade | Chemical Solution |
| Packaging | Poly Bottle |
| Color | Colorless |
| Quantity | 500 mL |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.