missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein, 1% (w/v) Indicator Solution in 95% Isopropyl Alcohol, Spectrum™ Chemical

Catalog No. 18612267
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18612267 125 mL
18612268 500 mL
18612269 1 L
18612270 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18612267 Supplier Spectrum Chemical Mfg Cor Supplier No. P155125ML

Specifications

Chemical Name or Material Phenolphthalein
CAS 77-09-8, 67-63-0, 7732-18-5
Concentration 1 Percent (w/v) Indicator Solution in 95 Percent Isopropyl Alcohol
Grade Chemical Solution
Packaging Poly Bottle
Color Colorless
Quantity 125 mL

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.