missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenolphthalein, 1% (w/v) in 50% (v/v) Isopropyl Alcohol, Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Phenolphthalein |
| CAS | 7732-18-5, 67-63-0, 77-09-8 |
| CAS Min % | 49.0, 0.5 |
| CAS Max % | 51.0, 1.5 |
| Packaging | Natural Poly Bottle |
| Color | Colorless to Pink |
| Quantity | 60 mL |
| Physical Form | Liquid |
| Percent Purity | 1% W/V |
| Grade | Indicator |