missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenolphthalein (1% in ca. 85% Ethanol) [for Titration], TCI America™
Specifications
Specifications
| UN Number | 1170 |
| Color | Colorless |
| Quantity | 100 mL |
| Physical Form | Liquid |
| Formula Weight | 318.33 |
| Chemical Name or Material | Phenolphthalein (1% in ca. 85% Ethanol) [for Titration] |
Safety and Handling
| EINECSNumber | (9)-1152 |
| RTECSNumber | SM8380000 |
| TSCA | Yes |