missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenolphthalein, 1% in 95% Ethanol, pH 8.3 to 10.0 Colorless to Pink, Certified, LabChem™
Specifications
Specifications
| Chemical Name or Material | Phenolphthalein |
| Name Note | 1% in 95% Ethanol |
| CAS Max % | 89.1, 4.95, 4.95, 1 |
| CAS | 67-63-0, 67-56-1, 77-09-8 |
| Linear Formula | C20H14O4 |
| Solubility Information | Soluble in water |
| Identification | Passes Test |
| Formula Weight | 318.32 |
| Grade | Certified |
| Packaging | Poly Bottle |
| Show More |