missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein (0.1% in ca. 90% Ethanol) [for Titration], TCI America™

Catalog No. P0701100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0701100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0701100ML Supplier TCI America Supplier No. P0701100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
Synonym phenolphthalein, phthalimetten, euchessina, phthalin, espotabs, phenolax, purgophen, koprol, laxogen, trilax
PubChem CID 4764
ChEBI CHEBI:34914
IUPAC Name 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Color Colorless
Quantity 100 mL
UN Number 1170
Formula Weight 318.33
Physical Form Liquid
Chemical Name or Material Phenolphthalein (0.1% in ca. 90% Ethanol) [for Titration]
EINECSNumber (9)-1152
RTECSNumber SM8380000
TSCA Yes