missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenolphthalein (0.04% in ca. 40% Ethanol) [for pH Determination], TCI America™
$36.07 - $68.34
Chemical Identifiers
| CAS | 77-09-8 |
|---|---|
| Molecular Formula | C20H14O4 |
| Molecular Weight (g/mol) | 318.33 |
| MDL Number | MFCD00005913 |
| InChI Key | KJFMBFZCATUALV-UHFFFAOYSA-N |
| PubChem CID | 4764 |
| ChEBI | CHEBI:34914 |
| IUPAC Name | 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one |
| SMILES | OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P0702100ML
|
TCI America
P0702100ML |
100 mL |
N/A
|
|
|||||
|
P0702500ML
|
TCI America
P0702500ML |
500 mL |
N/A
|
|
|||||
Chemical Identifiers
| 77-09-8 | |
| 318.33 | |
| KJFMBFZCATUALV-UHFFFAOYSA-N | |
| CHEBI:34914 | |
| OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |
| C20H14O4 | |
| MFCD00005913 | |
| 4764 | |
| 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one |
Specifications
| 77-09-8 | |
| MFCD00005913 | |
| KJFMBFZCATUALV-UHFFFAOYSA-N | |
| 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one | |
| 4764 | |
| 318.33 | |
| 100 mL | |
| Phenolphthalein (0.04% in ca. 40% Ethanol) [for pH Determination] |
| C20H14O4 | |
| 1170 | |
| OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 | |
| 318.33 | |
| CHEBI:34914 | |
| Colorless | |
| Liquid |
Safety and Handling
EINECSNumber : (9)-1152
RTECSNumber : SM8380000
TSCA : Yes