missing translation for 'onlineSavingsMsg'
Learn More

Phenolphthalein (0.04% in ca. 40% Ethanol) [for pH Determination], TCI America™

Catalog No. P0702100ML
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0702100ML 100 mL
P0702500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0702100ML Supplier TCI America Supplier No. P0702100ML
Only null left

Chemical Identifiers

CAS 77-09-8
Molecular Formula C20H14O4
Molecular Weight (g/mol) 318.33
MDL Number MFCD00005913
InChI Key KJFMBFZCATUALV-UHFFFAOYSA-N
PubChem CID 4764
ChEBI CHEBI:34914
IUPAC Name 3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
SMILES OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

UN Number 1170
Formula Weight 318.33
Color Colorless
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Phenolphthalein (0.04% in ca. 40% Ethanol) [for pH Determination]
EINECSNumber (9)-1152
RTECSNumber SM8380000
TSCA Yes