Learn More
Qorpak™ Phenolic Pulp/Vinyl Lined Caps
Catalog No. 02913383
Change view
Click to view available options
Thread Size (Metric):
AP mm
Catalog No. Thread Size (Metric)
02913383 AP mm
1 options
Catalog No. 02913383 Supplier Qorpak™ Supplier No. CAP00089
Only null left
This general purpose liner is commonly used for food, beverage, medical and chemical applications
  • For general use
  • The liner is made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
  • This general purpose liner is commonly used for:
    • Food
    • Beverage
    • Medical and chemical applications packed at less than 120° F (including mild acids, alkalis, solvents, alcohols, oils and aqueous products; NOT recommended for active hydrocarbons or bleaches)
Specifications
Color Black
Material Phenolic Resin
No. per Case 240
Packaging Bulk
Sterility Non-sterile
Thread Size (Metric) AP mm
Liner Material Pulp/Vinyl
Neck Finish 89-400
Disposable Reusable
Product Type Threaded Screw Cap
Diameter (Metric) 89 mm
Autoclavable Not Autoclavable
Show More Show Less