missing translation for 'onlineSavingsMsg'
Learn More

pH 4.7 Phenol Red TS, Ricca Chemical

$117.22 - $233.34

Specifications

Chemical Name or Material pH 4.7 Phenol Red
Name Note Test Solution
CAS 7732-18-5,64-19-7,1310-73-2,7783-20-2,143-74-8,7783-20-2
CAS Min % 92.0,3.08,2.97,0.06,0.02,0.0045
CAS Max % 96.0,3.2,3.09,0.06,0.02,0.0047
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
5721-4
View Documents
Ricca Chemical Company
57214
120 mL Natural Poly Bottle
Each for $117.22
Only null left
 
5721-16
View Documents
Ricca Chemical Company
572116
500 mL Natural Poly Bottle
Each for $233.34
Only null left
 
Description

Description

Specifications

Specifications

pH 4.7 Phenol Red
7732-18-5,64-19-7,1310-73-2,7783-20-2,143-74-8,7783-20-2
96.0,3.2,3.09,0.06,0.02,0.0047
Natural Poly Bottle
4.7
Test Solution
92.0,3.08,2.97,0.06,0.02,0.0045
Indicator
Yellow
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'