missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red, TCI America™

Catalog No. P01001G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P01001G 1 g
P010025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P01001G Supplier TCI America Supplier No. P01001G

Chemical Identifiers

CAS 143-74-8
Molecular Formula C19H14O5S
Molecular Weight (g/mol) 354.38
MDL Number MFCD00003552
InChI Key BELBBZDIHDAJOR-UHFFFAOYSA-N
Synonym Phenolsulfonphthalein
PubChem CID 4766
ChEBI CHEBI:31991
IUPAC Name 3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
SMILES OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Formula Weight 354.38
Quantity 1 g
Physical Form Crystal/Powder
Chemical Name or Material Phenol Red
EINECSNumber (4)-0271
RTECSNumber SJ7490000
TSCA Yes