missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red, Sodium Salt (Certified ACS), Fisher Chemical

Catalog No. P39110
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P39110 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P39110 Supplier Thermo Fisher Scientific Supplier No. P391 10
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

pH indicator

Chemical Identifiers

CAS 34487-61-1
Molecular Formula C19H13NaO5S
Molecular Weight (g/mol) 376.358
MDL Number MFCD00066901
InChI Key HKHYOKBQJILTEI-UHFFFAOYSA-M
Synonym Phenolsulfonephthalein Sodium Salt
PubChem CID 23686673
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]

Specifications

Melting Point 285°C
Clarity Pass Test
Identification Pass Test
Packaging Glass Bottle
Visual Transition Interval Pass Test
Linear Formula C6H4SO2OC(C6H4-4-OH)(C6H4-4-ONa)
Decomposition Information Decomposition Information: 285°C
Color Brown/Red
Quantity 10 g
Physical Form Powder
Formula Weight 376.37
Grade Certified ACS
Chemical Name or Material Phenol Red, Sodium Salt
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category SIGNAL WORD: CAUTION!
Precautionary Statement Emergency Overview
Causes eye, skin, and respiratory tract irritation.
NFPA
Health: 2
Flammability: 1
Instability: 0