missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenol Red sodium salt, ACS reagent

CAS: 34487-61-1 | C19H13NaO5S | 376.358 g/mol

$58.50 - $457.72

Chemical Identifiers

CAS 34487-61-1
Molecular Formula C19H13NaO5S
Molecular Weight (g/mol) 376.358
MDL Number MFCD00066901
InChI Key HKHYOKBQJILTEI-UHFFFAOYSA-M
PubChem CID 23686673
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC417250050
SDS View Documents
Thermo Scientific Chemicals
417250050
5 g Glass bottle
Each for $58.50
Only null left
 
AC417250100
SDS View Documents
Thermo Scientific Chemicals
417250100
10 g Glass bottle
Each for $103.24
Case of 6 Each for $0.00
Only null left
 
AC417251000
SDS View Documents
Thermo Scientific Chemicals
417251000
100 g Glass bottle
Each for $457.72
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

34487-61-1
376.358
HKHYOKBQJILTEI-UHFFFAOYSA-M
sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
C19H13NaO5S
MFCD00066901
23686673
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]

Specifications

285.0°C
passes test
MFCD00066901
15, 7360
HKHYOKBQJILTEI-UHFFFAOYSA-M
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]
376.358
376.35
Glass bottle
White
Crystalline Powder or Flakes
34487-61-1
C19H13NaO5S
19, 91
Solubility in water: soluble.
Authentic
sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
23686673
ACS Reagent
from pH 6.8 (yellow) to pH 8.2 (red)
5 g
Phenol Red sodium salt
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents

RUO – Research Use Only