Learn More
Thermo Scientific Chemicals Phenol Red sodium salt, ACS
Description
Phenol red sodium salt is a pH indicator commonly used in cell culture for monitoring pH changes. It is used in the pH ranges between 6.8 and 8.2. It is involved in the studies of gastrointestinal and other systems. It can be used to check hydrogen peroxide in supernatants from cultured macrophages in multiwell plates. Further, it is used as a diagnostic aid for determination of renal function.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 285°C (decomposition) |
| Beilstein | 6260026 |
| Merck Index | 14,7247 |
| Solubility Information | Soluble in water,alcohol,aqueous alkali hydroxides and carbonates. |
| Quantity | 25 g |
| Physical Form | Crystalline |
| Formula Weight | 376.37 |
| Grade | ACS |
| Chemical Name or Material | Phenol Red sodium salt |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 252-057-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only