Learn More
Thermo Scientific Chemicals Phenol Red sodium salt
CAS: 34487-61-1 | C19H13NaO5S | 376.358 g/mol
Supplier: Thermo Scientific Chemicals B2194814
Description
Phenol Red sodium salt commonly employed in cell biology laboratories as a cell tissue culture pH marker. Under normal conditions, phenol red added to the growth medium will have a pink-red color. If their is dying cells or overgrowth of contaminants, this will cause a change in pH resulting in a change in indicator color. This color change is an indication that the medium needs to be replaced. Additionally, Phenol Red could potentially be useful as a differentiation factor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenol Red sodium salt commonly employed in cell biology laboratories as a cell tissue culture pH marker. Under normal conditions, phenol red added to the growth medium will have a pink-red color. If their is dying cells or overgrowth of contaminants, this will cause a change in pH resulting in a change in indicator color. This color change is an indication that the medium needs to be replaced. Additionally, Phenol Red could potentially be useful as a differentiation factor.
Solubility
Soluble in water yielding a dark red solution (25 mg/ml), alcohol (4 mg/ml), methanol, ethanol (200 mg/ml), and aqueous alkali hydroxides or carbonates with formation of a red colored solution.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
| Phenol Red sodium salt | |
| 285°C (decomposition) | |
| MFCD00066901 | |
| 6260026 | |
| phenol red, sodium salt, phenolsulfonephthalein sodium salt, sodium 4-3-4-hydroxyphenyl-1,1-dioxido-3h-benzo c 1,2 oxathiol-3-yl phenolate, phenol red sodium salt, phenol, 4,4'-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis-, monosodium salt, phenol, 4,4'-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis-, sodium salt 1:1, phenol red, acs, phenolredsodiumsalt, phenolsulfonephthalein sodium, sodium 4-3-4-hydroxyphenyl-1,1-dioxo-2,1??-benzoxathiol-3-yl benzenolate | |
| HKHYOKBQJILTEI-UHFFFAOYSA-M | |
| sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate | |
| 23686673 |
| 34487-61-1 | |
| C19H13NaO5S | |
| 25 g | |
| 14,7247 | |
| Soluble in water yielding a dark red solution (25mg/ml),alcohol (4mg/ml),methanol,ethanol (200mg/ml),and aqueous alkali hydroxides or carbonates with formation of a red colored solution. | |
| C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+] | |
| 376.358 | |
| 376.37 |
Chemical Identifiers
| 34487-61-1 | |
| 376.358 | |
| HKHYOKBQJILTEI-UHFFFAOYSA-M | |
| 23686673 | |
| C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+] |
| C19H13NaO5S | |
| MFCD00066901 | |
| phenol red, sodium salt, phenolsulfonephthalein sodium salt, sodium 4-3-4-hydroxyphenyl-1,1-dioxido-3h-benzo c 1,2 oxathiol-3-yl phenolate, phenol red sodium salt, phenol, 4,4'-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis-, monosodium salt, phenol, 4,4'-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis-, sodium salt 1:1, phenol red, acs, phenolredsodiumsalt, phenolsulfonephthalein sodium, sodium 4-3-4-hydroxyphenyl-1,1-dioxo-2,1??-benzoxathiol-3-yl benzenolate | |
| sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 252-057-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only