missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Phenol Red, sodium salt, pure, water soluble, indicator
CAS: 34487-61-1 | C19H13NaO5S | 376.358 g/mol
$53.15 - $165.53
Chemical Identifiers
| CAS | 34487-61-1 |
|---|---|
| Molecular Formula | C19H13NaO5S |
| Molecular Weight (g/mol) | 376.358 |
| InChI Key | HKHYOKBQJILTEI-UHFFFAOYSA-M |
| PubChem CID | 23686673 |
| IUPAC Name | sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate |
| SMILES | C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC151440050
|
Thermo Scientific Chemicals
151440050 |
5 g | Glass bottle |
N/A
|
|
||||
|
AC151440250
|
Thermo Scientific Chemicals
151440250 |
25 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
| 34487-61-1 | |
| 376.358 | |
| 23686673 | |
| C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+] |
| C19H13NaO5S | |
| HKHYOKBQJILTEI-UHFFFAOYSA-M | |
| sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate |
Specifications
| 285.0°C | |
| C19H13NaO5S | |
| HKHYOKBQJILTEI-UHFFFAOYSA-M | |
| C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+] | |
| 376.358 | |
| 376.35 | |
| Glass bottle | |
| pH 6.8 (yellow) to pH 8.2 (red) | |
| 5 g | |
| Phenol Red, sodium salt |
| 34487-61-1 | |
| Solubility in water: soluble. | |
| Authentic | |
| sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate | |
| 23686673 | |
| Pure | |
| 1600 (558nm, in buffer) min. | |
| Brown to Red | |
| Amorphous Powder |
Safety and Handling
Warning
RUO – Research Use Only