missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenol Red, sodium salt, pure, water soluble, indicator

Catalog No. AC151440050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC151440050 5 g Glass bottle
AC151440250 25 g Glass bottle
AC151441000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC151440050 Supplier Thermo Scientific Chemicals Supplier No. 151440050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 34487-61-1 | C19H13NaO5S | 376.358 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 34487-61-1
Molecular Formula C19H13NaO5S
Molecular Weight (g/mol) 376.358
InChI Key HKHYOKBQJILTEI-UHFFFAOYSA-M
PubChem CID 23686673
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]

Specifications

Melting Point 285.0°C
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Formula Weight 376.35
Grade Pure
Packaging Glass bottle
Specific Extinction 1600 (558nm, in buffer) min.
Visual Transition Interval pH 6.8 (yellow) to pH 8.2 (red)
Color Brown to Red
Quantity 5 g
Physical Form Amorphous Powder
Chemical Name or Material Phenol Red, sodium salt
Show More Show Less
Hazard Category Warning

RUO – Research Use Only