missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red sodium salt, 0.02% w/v aq. soln.

CAS: 34487-61-1 | C19H13NaO5S | 376.358 g/mol

$73.99 - $73.99

Chemical Identifiers

CAS 34487-61-1
Molecular Formula C19H13NaO5S
Molecular Weight (g/mol) 376.358
MDL Number MFCD00066901
InChI Key HKHYOKBQJILTEI-UHFFFAOYSA-M
PubChem CID 23686673
IUPAC Name sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
SMILES C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA38702AP
View Documents
Thermo Scientific Chemicals
038702AP
500 mL
Each for $73.99
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

34487-61-1
376.358
HKHYOKBQJILTEI-UHFFFAOYSA-M
sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
C19H13NaO5S
MFCD00066901
23686673
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]

Specifications

34487-61-1
C19H13NaO5S
HKHYOKBQJILTEI-UHFFFAOYSA-M
sodium;4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenolate
23686673
0.972 g/mL
Liquid
0.02% w/v aq. soln.
MFCD00066901
C1=CC=C2C(=C1)C(OS2(=O)=O)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)[O-].[Na+]
376.358
376.37
500 mL
Phenol Red sodium salt
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 252-057-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only