missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red Indicator, ACS Reagent, Solstice

Catalog No. 60046147
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046147 25 g Glass Bottle
60046146 10 g Glass Bottle
60046145 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60046147 Supplier Solstice Supplier No. 3266125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Indicator, ACS reagent

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 143-74-8
Molecular Formula C19H14O5S
Molecular Weight (g/mol) 354.38
MDL Number MFCD00003552
InChI Key BELBBZDIHDAJOR-UHFFFAOYSA-N
Synonym Phenolsulfonphthalein
PubChem CID 4766
ChEBI CHEBI:31991
IUPAC Name 4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Packaging Glass Bottle
Visual Transition Interval 6.8 to 8.2 (yellow to red)
UN Number NONH for all modes of transport
Beilstein 326470
Color Red
Quantity 25 g
Physical Form Powder
Formula Weight 354.38g/mol
Grade ACS Reagent
Chemical Name or Material Phenol Red
Hazard Statement P261-P305 + P351 + P338
Precautionary Statement H315-H319-H335
EINECSNumber 205-609-7
RTECSNumber SJ7490000