missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red Indicator, 0.1% w/v, Reagents

Catalog No. 01334378
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334378 500 mL
01334380 1 L
01334379 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01334378 Supplier Labchem Inc Supplier No. CP109300500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phenol Red Indicator, 0.1% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Phenol Red Indicator, 0.1% w/v
CAS 7732-18-5, 34487-61-1
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.0003
Color Orange/Red
Synonym Phenolsulfonphthalein
Quantity 500 mL
Physical Form Liquid
Percent Purity 0.1%
Grade Reagent
Show More Show Less