missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol Red, ACS Reagent (Phenolsulfonphthalein), Reagents
Phenol Red, ACS Reagent (Phenolsulfonphthalein), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
$162.18 - $333.71
Chemical Identifiers
| CAS | 143-74-8 |
|---|---|
| Molecular Formula | C19H14O5S |
| Molecular Weight (g/mol) | 354.38 |
| InChI Key | BELBBZDIHDAJOR-UHFFFAOYSA-N |
| Synonym | Phenolsulfonphthalein |
| IUPAC Name | 3,3-bis(4-hydroxyphenyl)-3H-2,1λâ¶-benzoxathiole-1,1-dione |
| SMILES | OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-336-732
|
Labchem Inc
C61225001C2 |
1 g |
Each for $162.18
|
|
|||||
|
01-336-731
|
Labchem Inc
C61225005C2 |
5 g |
Each for $333.71
|
|
|||||
Chemical Identifiers
| 143-74-8 | |
| 354.38 | |
| Phenolsulfonphthalein | |
| OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |
| C19H14O5S | |
| BELBBZDIHDAJOR-UHFFFAOYSA-N | |
| 3,3-bis(4-hydroxyphenyl)-3H-2,1λâ¶-benzoxathiole-1,1-dione |
Specifications
| 563°C | |
| C19H14O5S | |
| BELBBZDIHDAJOR-UHFFFAOYSA-N | |
| 3,3-bis(4-hydroxyphenyl)-3H-2,1λâ¶-benzoxathiole-1,1-dione | |
| ACS | |
| 1 | |
| 1 g | |
| Phenol Red, ACS Reagent (Phenolsulfonphthalein) |
| 143-74-8 | |
| Phenolsulfonphthalein | |
| OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 | |
| 354.38 | |
| Glass Bottle | |
| Red | |
| Solid |