missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol Red, ACS Reagent (Phenolsulfonphthalein), Reagents
Specifications
Specifications
| Boiling Point | 563°C |
| Packaging | Glass Bottle |
| Specific Gravity | 1 |
| Color | Red |
| Quantity | 1 g |
| Physical Form | Solid |
| Grade | ACS |
| Chemical Name or Material | Phenol Red, ACS Reagent (Phenolsulfonphthalein) |