Learn More
Thermo Scientific Chemicals Phenol Red
CAS: 143-74-8 | C19H14O5S | 354.38 g/mol
Supplier: Thermo Scientific Chemicals B2171009
Description
Phenol red is used as a non absorbed marker in gravimetric method. It is a pH indicator used in cell biology laboratories and in home swimming pool test kits. It is used to check the kidney function based on the excreted phenol red by colorimetric method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhenol red is used as a non absorbed marker in gravimetric method. It is a pH indicator used in cell biology laboratories and in home swimming pool test kits. It is used to check the kidney function based on the excreted phenol red by colorimetric method.
Solubility
Soluble in water, ethanol and methanol. Insoluble in chloroform.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Phenol Red | |
| >300°C | |
| C19H14O5S | |
| 10 g | |
| 14,7247 | |
| Soluble in water,ethanol and methanol. Insoluble in chloroform. | |
| OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 | |
| 354.38 | |
| CHEBI:31991 |
| 143-74-8 | |
| Odorless | |
| MFCD00003552 | |
| 326470 | |
| phenol red, phenolsulfonphthalein, fenolipuna, sulfonphthal, sulphental, sulphonthal, phenolsulphonphthalein, psp indicator, phenolsulfonephthalein, unii-i6g9y0j1oj | |
| BELBBZDIHDAJOR-UHFFFAOYSA-N | |
| 4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol | |
| 4766 | |
| 354.38 |
Chemical Identifiers
| 143-74-8 | |
| 354.38 | |
| BELBBZDIHDAJOR-UHFFFAOYSA-N | |
| 4766 | |
| OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1 |
| C19H14O5S | |
| MFCD00003552 | |
| phenol red, phenolsulfonphthalein, fenolipuna, sulfonphthal, sulphental, sulphonthal, phenolsulphonphthalein, psp indicator, phenolsulfonephthalein, unii-i6g9y0j1oj | |
| CHEBI:31991 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 205-609-7
RTECSNumber : SJ7490000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only