missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenol Red

Catalog No. AAB2171030
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2171030 250 g
AAB2171009 10 g
AAB2171018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2171030 Supplier Thermo Scientific Chemicals Supplier No. B2171030

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 143-74-8 | C19H14O5S | 354.38 g/mol

Phenol red is used as a non absorbed marker in gravimetric method. It is a pH indicator used in cell biology laboratories and in home swimming pool test kits. It is used to check the kidney function based on the excreted phenol red by colorimetric method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenol red is used as a non absorbed marker in gravimetric method. It is a pH indicator used in cell biology laboratories and in home swimming pool test kits. It is used to check the kidney function based on the excreted phenol red by colorimetric method.

Solubility
Soluble in water, ethanol and methanol. Insoluble in chloroform.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 143-74-8
Molecular Formula C19H14O5S
Molecular Weight (g/mol) 354.38
MDL Number MFCD00003552
InChI Key BELBBZDIHDAJOR-UHFFFAOYSA-N
Synonym phenol red, phenolsulfonphthalein, fenolipuna, sulfonphthal, sulphental, sulphonthal, phenolsulphonphthalein, psp indicator, phenolsulfonephthalein, unii-i6g9y0j1oj
PubChem CID 4766
ChEBI CHEBI:31991
SMILES OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Boiling Point >300°C
Odor Odorless
Quantity 250 g
Beilstein 326470
Merck Index 14,7247
Solubility Information Soluble in water,ethanol and methanol. Insoluble in chloroform.
IUPAC Name 3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Formula Weight 354.38
Chemical Name or Material Phenol Red
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-609-7
RTECSNumber SJ7490000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only