missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol Red, 0.5% Solution, MP Biomedicals
Description
- Phenol Red is used as a pH indicator in cell culture applications
- A solution of phenol red will have a yellow color at a pH of 6.4 or below and a red color at a pH of 8.2 and above
- Phenol red in tissue culture media can act as a weak estrogen, especially with human breast cancer cells
- Phenol red can be used to measure hydrogen peroxide in cultured macrophages in multiwell plates
Specifications
Specifications
| Linear Formula | C19H13O5SNa(Lit.) |
| Color | Orange |
| Solubility Information | Soluble in water (100mg/mL-clear red solution) methanol or ethanol (200mg/mL). |
| Quantity | 100 mL |
| Physical Form | Solution |
| Formula Weight | 376.36(Lit.) |
| Chemical Name or Material | PHENOL RED, 0.5% Solution |
Safety and Handling
| Recommended Storage | Store at Room Temperature (15-30°C). |