missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red (0.1% in ca. 20% Ethanol) [for Titration], TCI America™

Catalog No. P0703500ML
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0703500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0703500ML Supplier TCI America Supplier No. P0703500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 143-74-8
Molecular Formula C19H14O5S
Molecular Weight (g/mol) 354.38
MDL Number MFCD00003552
InChI Key BELBBZDIHDAJOR-UHFFFAOYSA-N
Synonym phenol red, phenolsulfonphthalein, fenolipuna, sulfonphthal, sulphental, sulphonthal, phenolsulphonphthalein, psp indicator, phenolsulfonephthalein, unii-i6g9y0j1oj
PubChem CID 4766
ChEBI CHEBI:31991
IUPAC Name 3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
SMILES OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

Color Red-Yellow
Quantity 500 mL
Formula Weight 354.38
Physical Form Liquid
Chemical Name or Material Phenol Red (0.1% in ca. 20% Ethanol) [for Titration]
EINECSNumber (4)-0271
RTECSNumber SJ7490000
TSCA Yes