missing translation for 'onlineSavingsMsg'
Learn More

Phenethylmagnesium chloride, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

$131.33 - $131.33

Chemical Identifiers

CAS 90878-19-6
Molecular Formula C8H9ClMg
Molecular Weight (g/mol) 164.92
MDL Number MFCD00192387
InChI Key AZORNQXQALOFHH-UHFFFAOYSA-M
Synonym phenethyl magnesium chloride, phenylethyl magnesium chloride, chloro 2-phenylethyl magnesium, phenethylmagnesium chloride, phenethylmagnesium chloride solution, phenethymagnesium chloride, phenyethylmagnesium chloride, phenylethylmagnesium chloride, 2-phenylethylmagnesium chloride, 2-phenylethyl magnesium chloride
PubChem CID 4671868
IUPAC Name magnesium;ethylbenzene;chloride
SMILES Cl[Mg]CCC1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC427801000
SDS View Documents
Thermo Scientific Chemicals
427801000
100 mL AcroSeal™ Glass bottle
Each for $131.33
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

90878-19-6
164.92
AZORNQXQALOFHH-UHFFFAOYSA-M
4671868
Cl[Mg]CCC1=CC=CC=C1
C8H9ClMg
MFCD00192387
phenethyl magnesium chloride, phenylethyl magnesium chloride, chloro 2-phenylethyl magnesium, phenethylmagnesium chloride, phenethylmagnesium chloride solution, phenethymagnesium chloride, phenyethylmagnesium chloride, phenylethylmagnesium chloride, 2-phenylethylmagnesium chloride, 2-phenylethyl magnesium chloride
magnesium;ethylbenzene;chloride

Specifications

90878-19-6,109-99-9
0.9540g/mL
AcroSeal™ Glass bottle
C6H5(CH2)2MgCl
100 mL
phenethyl magnesium chloride, phenylethyl magnesium chloride, chloro 2-phenylethyl magnesium, phenethylmagnesium chloride, phenethylmagnesium chloride solution, phenethymagnesium chloride, phenyethylmagnesium chloride, phenylethylmagnesium chloride, 2-phenylethylmagnesium chloride, 2-phenylethyl magnesium chloride
Cl[Mg]CCC1=CC=CC=C1
164.92
164.92
Phenethylmagnesium chloride, 1M solution in THF
Yellow to Brown
−17°C
C8H9ClMg
MFCD00192387
0.954
AZORNQXQALOFHH-UHFFFAOYSA-M
magnesium;ethylbenzene;chloride
4671868
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only