missing translation for 'onlineSavingsMsg'
Learn More

Phenethylamine, 99%

Catalog No. AC156492500
Change view
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156492500 250 g Glass bottle
AC156491000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC156492500 Supplier Thermo Scientific Chemicals Supplier No. 156492500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 64-04-0 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64-04-0
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00008184
InChI Key BHHGXPLMPWCGHP-UHFFFAOYSA-N
Synonym phenethylamine, 2-phenylethylamine, benzeneethanamine, phenylethylamine, 2-phenethylamine, beta-phenylethylamine, beta-phenethylamine, 1-amino-2-phenylethane, 2-aminoethyl benzene, beta-aminoethylbenzene
PubChem CID 1001
ChEBI CHEBI:18397
IUPAC Name 2-phenylethanamine
SMILES C1=CC=C(C=C1)CCN

Specifications

Melting Point -60.0°C
Color Colorless to Yellow
Density 0.9850g/mL
Boiling Point 200.0°C to 202.0°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5320 to 1.5340
Linear Formula C6H5CH2CH2NH2
Quantity 250 g
Beilstein 12, 1096
Fieser 09,363
Specific Gravity 0.985
Merck Index 15, 7339
Solubility Information Solubility in water: 63g/L (25°C). Other solubilities: soluble in ethanol, diethyl ether
Viscosity 2.4 mm2/s (23°C)
Formula Weight 121.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Phenethylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 200-574-4

RUO – Research Use Only