missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenethyl Isocyanate 98.0+%, TCI America™
Supplier: TCI America P167725G
Specifications
| Phenethyl Isocyanate | |
| Yellow | |
| C9H9NO | |
| 25 g | |
| phenethyl isocyanate, 2-isocyanatoethyl benzene, phenylethyl isocyanate, 2-phenylethyl isocyanate, 2-phenylethylisocyanate, phenylethylisocyanate, phenethylisocyanate, benzene, 2-isocyanatoethyl, 2-isocyanato-ethyl-benzene, 2-phenethyl isocyanate | |
| C1=CC=C(C=C1)CCN=C=O | |
| 147.177 | |
| CHEBI:60067 | |
| ≥98.0% (GC) |
| 1943-82-4 | |
| 210°C | |
| MFCD00037044 | |
| 2206 | |
| HACRKYQRZABURO-UHFFFAOYSA-N | |
| 2-isocyanatoethylbenzene | |
| 160602 | |
| 147.18 | |
| Liquid |
Chemical Identifiers
| 1943-82-4 | |
| 147.177 | |
| HACRKYQRZABURO-UHFFFAOYSA-N | |
| 160602 | |
| 2-isocyanatoethylbenzene |
| C9H9NO | |
| MFCD00037044 | |
| phenethyl isocyanate, 2-isocyanatoethyl benzene, phenylethyl isocyanate, 2-phenylethyl isocyanate, 2-phenylethylisocyanate, phenylethylisocyanate, phenethylisocyanate, benzene, 2-isocyanatoethyl, 2-isocyanato-ethyl-benzene, 2-phenethyl isocyanate | |
| CHEBI:60067 | |
| C1=CC=C(C=C1)CCN=C=O |
Safety and Handling
TSCA : No